Dihydrofuran-3(2H)-one ≥98.0%

Supplier: TCI America

T2971-1G T2971-5G
TCT2971-1GEA 41.8 USD
TCT2971-1G TCT2971-5G
Dihydrofuran-3(2H)-one ≥98.0%
Dihydrofuran-3(2H)-one

CAS Number: 22929-52-8
MDL Number: MFCD07778393
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 68
Flash Point (°C): 56
Specific Gravity (20/20): 1.12

Formula: C₄H₆O₂
Boiling Pt: 68 °C
Density: 1.12 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD07778393
CAS Number: 22929-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c96c2d60729449849b97e0dc7aadefc491b9e889&vid=c96c2d60729449849b97e0dc7aadefc491b9e889&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR