1-Hydroxy-6-(trifluoromethyl)benzotriazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H1454-1G H1454-5G
TCH1454-1GEA 65 USD
TCH1454-1G TCH1454-5G
1-Hydroxy-6-(trifluoromethyl)benzotriazole ≥98.0% (by HPLC, titration analysis)
1-Hydroxy-6-(trifluoromethyl)benzotriazole

CAS Number: 26198-21-0
MDL Number: MFCD00038806
Molecular Formula: C7H4F3N3O
Molecular Weight: 203.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 148
Lambda max.: 362 nm

Formula: C₇H₄F₃N₃O
MW: 203.12 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00038806
CAS Number: 26198-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b3e84b8110398763458925e520bedcdd84e8cfa&vid=0b3e84b8110398763458925e520bedcdd84e8cfa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR