3,5-Heptanedione ≥97.0%

Supplier: TCI America

H1395-25G H1395-5G
TCH1395-25GEA 220.72 USD
TCH1395-25G TCH1395-5G
3,5-Heptanedione ≥97.0%
3,5-Heptanedione

CAS Number: 7424-54-6
MDL Number: MFCD00015186
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 57
Specific Gravity (20/20): 0.95

Formula: C₇H₁₂O₂
Boiling Pt: 177 °C
Density: 0.95 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00015186
CAS Number: 7424-54-6
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b7a7fc667050bfaf5cf61210e729ed6a18350f&vid=21b7a7fc667050bfaf5cf61210e729ed6a18350f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR