1-Chloro-4-ethynylbenzene ≥98.0%

Supplier: TCI America

Synonyms: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne

C2670-1G C2670-5G
TCC2670-1GEA 78.5 USD
TCC2670-1G TCC2670-5G
1-Chloro-4-ethynylbenzene ≥98.0%
1-Chloro-4-ethynylbenzene

CAS Number: 873-73-4
MDL Number: MFCD00191917
Molecular Formula: C8H5Cl
Molecular Weight: 136.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 60
Melting point (°C): 47
Flash Point (°C): 57
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Chlorophenylacetylene

Formula: C₈H₅Cl
MW: 136.58 g/mol
Boiling Pt: 60 °C
Melting Pt: 47 °C
Flash Pt: 57 °C
MDL Number: MFCD00191917
CAS Number: 873-73-4
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec06d9f4734c1c89b3398cf863b76b68012d7716&vid=ec06d9f4734c1c89b3398cf863b76b68012d7716&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR