Bovine Casein (from Milk)
Supplier: MilliporeSigma
218680-25GM 218680-100GM
80108-888EA 93.92 USD
80108-888 80108-886
Bovine Casein (from Milk)
Proteins and Peptides
  • High molecular weight phosphoprotein
  • Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases