Fluoropolymer FEP Tubing, Norell
Supplier: Norell Inc
TWT-8 TWT-312 TWT-6 TWT-10 TWT-3 TWT-7 TWT-4 TWT-12 TWT-5 TWT-9
100554-338PK 102.45 USD
100554-338 100554-190 100554-154 100554-188 100554-152 100554-186 100554-322 100554-336 100554-306 100554-140
Fluoropolymer FEP Tubing, Norell
Tubing Transfer Tubing
Thin-wall transparent fluoropolymer FEP tubing is manufactured from special virgin grade DuPont thermoplastic FEP fluoropolymer.
  • Unlike PTFE tubing, this material can be heat sealed
Fluoropolymer tubing can also be used where snug and tight fit over glass or metal tubing is required.