Casein Peptone, HiMedia
Supplier: HiMedia
RM714-500G RM714-10KG RM714-2.5KG RM714-25KG RM714-5KG
61001-642EA 172.95 USD
61001-642 89229-968 89229-970 89229-972 89229-974
Casein Peptone, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Casein peptone is used to support the growth of fastidious microorganisms and is suitable for use in industrial fermentation.
  • Enzymic digest of milk protein
  • Nitrogen source for microorganisms
  • Suitable for use in industrial fermentation
HiCase Peptone is enzymic digest of milk protein. It is manufactured under controlled conditions. Milk protein being a rich source of amino nitrogen, the hydrolysed product HiCase Peptone obtained also contains peptides, aminoacids etc. which acts as nitrogen source for microorganisms when used in media formulations.