Petroleum spirit, 40…60 °C, CHROMASOLV™ for pesticide residue analysis, low boiling point hydrogen treated naphtha, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Petroleum ethers are low boiling aliphatic fractions, which are derived from crude petroleum via fractional distillation. The boiling range is 40 to 60 °C, which gives a high evaporation rate. Suitable for GC and Pesticide Residue Analysis.
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Boiling Pt:
40…60 °C (1.013 hPa) Density: 0.65 g/cm³ (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 309-852-0 UN: 3295 ADR: 3,II |
Specification Test Results
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.01 % |
Suitability for residue analysis | Max. 5 ng/l |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...