2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

34862-1L 34862-2.5L 34862-4X2.5L 34862P-4X4L
BJ34862-4LCS 1640.61 USD
BJ34862-4L BJ34862-1L BJ34862-2.5L BJ34862-4X2.5L BJ34862P-4X4L
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
2,2,4-Trimethylpentane

2,2,4-Trimethylpentane (C8H18), also know as Iso-Octane, generally serves as a nonpolar chomatography solvent. Suitable for HPLC and spectrophotometry.


  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Beilstein Number: 1696876

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2
Melting Pt: -107
Density: 0.69 g/cm³ (20 °C) (lit.)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.001 %
Absorbance at 270 nm Max. 0.01
Absorbance at 235 nm Max. 0.10
Absorbance at 210 nm Max. 0.70


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR