Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

34855-4X4L 34855-2.5L 34855-4X2.5L
BJ34855-4LCS 1327.25 USD
BJ34855-4L BJ34855-1L BJ34855-2.5L BJ34855-4X2.5L
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Cyclohexane

Cyclohexane (C6H12) is a saturated hydrocarbon and has a six member ring-structure. It is derived from the catalytic hydrogenation of benzene. Suitable for HPLC and spectrophotometry.


  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.78 g/cm³ (20 °C) (lit.)
Flash Pt: 8.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 99.70 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 255 nm Max. 0.01
Absorbance at 250 nm Max. 0.03
Absorbance at 240 nm Max. 0.08
Absorbance at 235 nm Max. 0.10
Absorbance at 230 nm Max. 0.20
Absorbance at 220 nm Max. 0.50
Absorbance at 210 nm Max. 1.00
Fluorescence (chinin) at 254 nm Max. 1.0 ppb
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
APHA Max. 10
Vapor Pressure 77 mmHg ( 20 °C)
Refractive Index n20/D 1.426 (lit.)
Explosion Limit 9 %
Autoignition Temperature 500 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR