Adhesive Vinyl Labels 'Sodium Hydroxide 1.00 Normal', HCL Labels
Supplier: HCL Label
GH1XX21VN0023 GH1XX21VN0035 GH1XX21VN0047 GH2XX21VN0023 GH2XX21VN0035 GH2XX21VN0047 GH3XX21VN0035 GH3XX21VN0047 GH3XX21VN0023 GH4XX21VN0023 GH4XX21VN0035 GH4XX21VN0047
103026-942PK 74.03 USD
103026-942 103026-944 103026-946 103026-948 103026-950 103026-952 103034-304 103034-306 103034-308 103034-310 103034-312 103034-314 103042-622 103042-624 103042-626 103042-628 103042-630 103042-980 103050-786 103050-788 103050-790 103050-792 103050-794 103050-796
Adhesive Vinyl Labels 'Sodium Hydroxide 1.00 Normal', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.