Adhesive Vinyl GHS OSHA Labels 'Cesium', HCL Labels
Supplier: HCL Label
GH12008VN0023 GH12008VN0035 GH12008VN0047 GH22008VN0023 GH22008VN0035 GH22008VN0047 GH32008VN0023 GH32008VN0035 GH32008VN0047 GH42008VN0023 GH42008VN0035 GH42008VN0047
103025-754PK 74.03 USD
103025-754 103025-756 103025-758 103025-760 103025-762 103025-764 103033-660 103033-662 103033-688 103033-690 103033-692 103033-694 103040-832 103040-834 103040-836 103040-838 103040-840 103040-842 103050-126 103050-128 103050-130 103050-132 103050-134 103050-136
Adhesive Vinyl GHS OSHA Labels 'Cesium', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labelling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.