Adhesive Vinyl GHS OSHA Labels 'Dlimonene', HCL Labels
Supplier: HCL Label
GH11988VN0023 GH11988VN0035 GH11988VN0047 GH21988VN0023 GH21988VN0035 GH21988VN0047 GH31988VN0035 GH31988VN0047 GH31988VN0023 GH41988VN0023 GH41988VN0035 GH41988VN0047
103025-662PK 74.03 USD
103025-662 103025-664 103025-666 103025-668 103025-670 103025-672 103033-592 103033-594 103033-596 103033-598 103033-600 103033-602 103040-806 103040-808 103040-810 103040-812 103040-814 103042-180 103049-264 103049-266 103049-268 103049-270 103049-834 103049-836
Adhesive Vinyl GHS OSHA Labels 'Dlimonene', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labelling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.