Adhesive Vinyl GHS OSHA Labels 'Isopropanol', HCL Labels
Supplier: HCL Label
GH11753VN0023 GH11753VN0035 GH11753VN0047 GH21753VN0023 GH21753VN0035 GH21753VN0047 GH31753VN0023 GH31753VN0035 GH31753VN0047 GH41753VN0023 GH41753VN0035 GH41753VN0047
103025-198PK 74.03 USD
103025-198 103025-200 103025-202 103025-204 103025-206 103025-208 103033-144 103033-146 103033-148 103033-150 103033-152 103033-154 103040-692 103041-606 103041-608 103041-610 103041-612 103041-646 103048-316 103049-116 103049-118 103049-120 103049-122 103049-124
Adhesive Vinyl GHS OSHA Labels 'Isopropanol', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.