Adhesive Vinyl GHS OSHA Labels 'Hydrofluoric Acid 50%', HCL Labels
Supplier: HCL Label
GH11391VN0023 GH11391VN0035 GH11391VN0047 GH21391VN0035 GH21391VN0047 GH21391VN0023 GH31391VN0047 GH31391VN0023 GH31391VN0035 GH41391VN0023 GH41391VN0035 GH41391VN0047
103024-402PK 74.03 USD
103024-402 103024-404 103024-406 103024-408 103024-562 103024-564 103032-296 103032-298 103032-300 103032-302 103032-508 103032-510 103040-206 103040-208 103040-210 103040-384 103040-386 103040-388 103047-744 103047-746 103047-748 103047-750 103047-752 103047-754
Adhesive Vinyl GHS OSHA Labels 'Hydrofluoric Acid 50%', HCL Labels
Labels
These ready-made laminated adhesive vinyl labels comply with the updated OSHA HazCom secondary container labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS). All sizes are protected with a chemical resistant lamination and a shelf/outdoor life of two years.
Certificaciones: RoHS compliant.