Adhesive Vinyl GHS OSHA Labels 'Sodium Thiosulfate', HCL Labels
Supplier: HCL Label
GH10620VN0023 GH10620VN0035 GH10620VN0047 GH20620VN0023 GH20620VN0035 GH20620VN0047 GH30620VN0023 GH30620VN0035 GH30620VN0047 GH40620VN0023 GH40620VN0035 GH40620VN0047
103023-390PK 74.03 USD
103023-390 103023-392 103023-394 103023-396 103023-398 103023-400 103031-288 103031-290 103031-292 103031-294 103031-296 103031-298 103039-188 103039-190 103039-192 103039-194 103039-196 103039-198 103046-740 103046-742 103046-744 103046-746 103046-748 103046-750
Adhesive Vinyl GHS OSHA Labels 'Sodium Thiosulfate', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labelling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.