Adhesive Vinyl GHS OSHA Labels 'Phenylethylamine', HCL Labels
Supplier: HCL Label
GH10575VN0023 GH10575VN00235 GH10575VN0035 GH10575VN00355 GH10575VN0047 GH10575VN00475 GH20575VN0023 GH20575VN00235 GH20575VN0035 GH20575VN00355 GH20575VN0047 GH20575VN00475 GH30575VN0023 GH30575VN00235 GH30575VN0035 GH30575VN00355 GH30575VN0047 GH30575VN00475 GH40575VN0023 GH40575VN00235 GH40575VN0035 GH40575VN00355 GH40575VN0047 GH40575VN00475
103022-910PK 74.03 USD
103022-910 103022-912 103022-914 103022-916 103022-918 103022-920 103030-736 103030-738 103030-740 103030-742 103030-744 103030-746 103038-104 103038-106 103038-108 103038-110 103038-112 103038-114 103048-762 103048-764 103048-766 103048-768 103048-770 103048-772
Adhesive Vinyl GHS OSHA Labels 'Phenylethylamine', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.