Adhesive Vinyl GHS OSHA Labels 'Periodic Acid', HCL Labels
Supplier: HCL Label
GH10574VN0023 GH10574VN0035 GH10574VN0047 GH20574VN0023 GH20574VN0035 GH20574VN0047 GH30574VN0023 GH30574VN0035 GH30574VN0047 GH40574VN0023 GH40574VN0035 GH40574VN0047
103022-730PK 74.03 USD
103022-730 103022-732 103022-902 103022-904 103022-906 103022-908 103030-724 103030-726 103030-728 103030-730 103030-732 103030-734 103038-044 103038-046 103038-096 103038-098 103038-100 103038-102 103048-750 103048-752 103048-754 103048-756 103048-758 103048-760
Adhesive Vinyl GHS OSHA Labels 'Periodic Acid', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.