Adhesive Vinyl GHS OSHA Labels 'Octane', HCL Labels
Supplier: HCL Label
GH10572VN0023 GH10572VN0035 GH10572VN0047 GH20572VN0023 GH20572VN0035 GH20572VN0047 GH30572VN0023 GH30572VN0035 GH30572VN0047 GH40572VN0023 GH40572VN0035 GH40572VN0047
103022-642PK 74.03 USD
103022-642 103022-644 103022-646 103022-648 103022-898 103022-900 103030-700 103030-702 103030-704 103030-706 103030-708 103030-710 103038-020 103038-022 103038-024 103038-026 103038-028 103038-030 103046-272 103046-274 103046-276 103046-278 103046-280 103046-282
Adhesive Vinyl GHS OSHA Labels 'Octane', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.