Adhesive Vinyl GHS OSHA Labels 'Magnesium', HCL Labels
Supplier: HCL Label
GH10540VN0023 GH10540VN0035 GH10540VN0047 GH20540VN0023 GH20540VN0035 GH20540VN0047 GH30540VN0023 GH30540VN0035 GH30540VN0047 GH40540VN0023 GH40540VN0035 GH40540VN0047
103022-090PK 74.03 USD
103022-090 103022-092 103022-094 103022-096 103022-098 103022-100 103030-332 103030-334 103030-336 103030-338 103030-340 103030-342 103037-996 103037-998 103038-440 103038-442 103038-444 103038-446 103045-936 103045-938 103045-940 103045-942 103045-944 103045-946
Adhesive Vinyl GHS OSHA Labels 'Magnesium', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant