Adhesive Vinyl GHS OSHA Labels 'Lead Acetate', HCL Labels
Supplier: HCL Label
GH10535VN0047 GH10535VN0023 GH10535VN0035 GH20535VN0023 GH20535VN0035 GH20535VN0047 GH30535VN0023 GH30535VN0035 GH30535VN0047 GH40535VN0023 GH40535VN0035 GH40535VN0047
103021-790PK 102.31 USD
103021-790 103021-792 103022-038 103022-040 103022-658 103022-660 103030-272 103030-274 103030-276 103030-278 103030-280 103030-282 103037-880 103037-882 103037-884 103037-886 103037-888 103037-890 103045-804 103045-806 103045-808 103045-810 103045-812 103045-814
Adhesive Vinyl GHS OSHA Labels 'Lead Acetate', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant