Adhesive Vinyl GHS OSHA Labels 'Iron Oxide', HCL Labels
Supplier: HCL Label
GH10530VN0023 GH10530VN0035 GH10530VN0047 GH20530VN0023 GH20530VN0035 GH20530VN0047 GH30530VN0035 GH30530VN0047 GH30530VN0023 GH40530VN0023 GH40530VN0035 GH40530VN0047
103021-978PK 74.03 USD
103021-978 103021-980 103021-982 103021-984 103021-986 103021-988 103029-516 103029-518 103029-520 103029-522 103029-524 103029-526 103037-848 103037-850 103037-852 103037-854 103038-420 103038-422 103045-864 103045-866 103045-868 103048-684 103048-686 103048-688
Adhesive Vinyl GHS OSHA Labels 'Iron Oxide', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant