Adhesive Vinyl GHS OSHA Labels 'Indium', HCL Labels
Supplier: HCL Label
GH10525VN0023 GH10525VN0035 GH10525VN0047 GH20525VN0023 GH20525VN0035 GH20525VN0047 GH30525VN0023 GH30525VN0035 GH30525VN0047 GH40525VN0023 GH40525VN0035 GH40525VN0047
103022-518PK 74.03 USD
103022-518 103022-520 103022-522 103022-524 103022-526 103022-528 103030-176 103030-178 103030-180 103030-182 103030-184 103030-186 103038-336 103038-338 103038-340 103038-342 103038-344 103038-346 103045-740 103045-742 103045-744 103045-746 103045-748 103045-750
Adhesive Vinyl GHS OSHA Labels 'Indium', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant