Adhesive Vinyl GHS OSHA Labels 'Boron Trichloride', HCL Labels
Supplier: HCL Label
GH10287VN0023 GH10287VN00235 GH10287VN0035 GH10287VN00355 GH10287VN0047 GH10287VN00475 GH20287VN0023 GH20287VN00235 GH20287VN0035 GH20287VN00355 GH20287VN0047 GH20287VN00475 GH30287VN0023 GH30287VN00235 GH30287VN0035 GH30287VN00355 GH30287VN0047 GH30287VN00475 GH40287VN0023 GH40287VN00235 GH40287VN0035 GH40287VN00355 GH40287VN0047 GH40287VN00475
103020-350PK 48.63 USD
103020-350 103020-352 103020-354 103020-356 103020-358 103020-360 103028-182 103028-184 103028-186 103028-188 103028-190 103028-192 103036-136 103036-138 103036-140 103036-142 103036-144 103036-146 103044-036 103044-038 103044-040 103044-042 103044-044 103044-046
Adhesive Vinyl GHS OSHA Labels 'Boron Trichloride', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.