Adhesive Vinyl GHS OSHA Labels 'Phosphoric Acid', HCL Labels
Supplier: HCL Label
GH10042VN0023 GH10042VN0035 GH10042VN0047 GH20042VN0023 GH20042VN0035 GH20042VN0047 GH30042VN0023 GH30042VN0035 GH30042VN0047 GH40042VN0023 GH40042VN0035 GH40042VN0047
103019-596PK 74.03 USD
103019-596 103019-598 103019-600 103019-602 103019-604 103019-606 103027-492 103027-494 103027-496 103027-498 103027-500 103027-502 103035-142 103035-144 103035-146 103035-148 103035-150 103035-152 103043-338 103043-340 103043-342 103043-344 103043-346 103043-348
Adhesive Vinyl GHS OSHA Labels 'Phosphoric Acid', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant