Adhesive Vinyl GHS OSHA Labels 'Carbon Tetrachloride', HCL Labels
Supplier: HCL Label
GH10029VN0023 GH10029VN0035 GH10029VN0047 GH20029VN0023 GH20029VN0035 GH20029VN0047 GH30029VN0023 GH30029VN0035 GH30029VN0047 GH40029VN0023 GH40029VN0035 GH40029VN0047
103019-408PK 74.03 USD
103019-408 103019-410 103019-412 103019-414 103019-416 103019-418 103027-320 103027-322 103027-324 103027-326 103027-328 103027-330 103034-994 103034-996 103034-998 103035-000 103035-002 103035-004 103043-190 103043-192 103043-194 103043-196 103043-198 103043-200
Adhesive Vinyl GHS OSHA Labels 'Carbon Tetrachloride', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant