Adhesive Vinyl GHS OSHA Labels 'Xylene', HCL Labels
Supplier: HCL Label
GH10012VN0023 GH10012VN0035 GH10012VN0047 GH20012VN0023 GH20012VN0035 GH20012VN0047 GH30012VN0023 GH30012VN0035 GH30012VN0047 GH40012VN0023 GH40012VN0035 GH40012VN0047
103019-254PK 74.03 USD
103019-254 103019-256 103019-258 103019-260 103019-262 103019-264 103027-198 103027-200 103027-202 103027-204 103027-206 103027-208 103034-904 103034-906 103034-908 103034-910 103034-912 103034-914 103043-108 103043-110 103043-112 103043-114 103043-116 103043-118
Adhesive Vinyl GHS OSHA Labels 'Xylene', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant