BioClean™ Legion™ Non sterile Latex Gloves, Ansell
Supplier: Ansell Healthcare
BLA301-S BLA302-M BLA303-L BLA304-XL
75834-008CS 795.22 USD
75834-008 75834-010 75834-012 75834-014
BioClean™ Legion™ Non sterile Latex Gloves, Ansell
Gloves
BioClean™ Legion™ non-sterile latex gloves offer unbeatable comfort with elbow length protection.
  • Exceptional flexibility and comfort
  • Elbow length
  • Powder-free
  • Beaded cuff
  • Textured for improved grip
  • Double-donnable
  • ISO Class 5 compatible
  • Non-particulating EasyTear™ packaging
For supreme comfort, exceptional flexibility, and elbow length protection, the 40.6 cm (16”) BioClean™ Legion™ Latex Glove is the optimal choice. With a textured surface for enhanced grip, this ambidextrous natural-colour glove is powder-free and contains a latex protein level of less than 50 μg/g.
Certificaciones: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 5 environment.
Complies with the requirements of Glove Standard EN 420:2003 and EN 374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN455-2:2009.
Envase: Gloves are packaged 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces).