NotI
Supplier: Promega Corporation
R6431
R6435
PAR6431EA
115.34
USD
PAR6431
PAR6435
NotI
Nucleic Acid Reagents
Restriction Enzymes
GC/GGCC GC CG CCGG\CG
- Capable of digesting DNA in 15 minutes or less
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
- High concentration product no longer available