SacI, Restriction Enzymes, Promega
Supplier: Promega Corporation
R6051 R6055
PAR6051EA 117.9 USD
PAR6051 PAR6055
SacI, Restriction Enzymes, Promega
Nucleic Acid Reagents Restriction Enzymes
G/TCGA C C AGCT\G
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications