Tissue Plasminogen Activator, Human, MilliporeSigma
Supplier: MilliporeSigma
612200-100UG
612200-1MG
80054-744EA
431.5
USD
80054-744
80054-746
Tissue Plasminogen Activator, Human, MilliporeSigma
Cells
Native tissue plasminogen activator from a human melanoma cell line.
- Prepared from cell line that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
- A serine protease that contains a single chain of 527 amino acids
- Binds to fibrin via lysine binding sites at its amino terminus and activates bound plasminogen
- Cleaves plasminogen to form active plasmin, the proteolytic enzyme that dissolves clots
- Lyophilized from 100 mM PBS, 3.5 mg/ml L-arginine, 0.001% Tween®-80 detergent
Purity: ≥95% by SDS-PAGE
Specific Activity: ≥300 KU/mg protein
Storage temperature: −20 °C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...