BBL™ Sabouraud Brain Heart Infusion Agar, Slants, BD
Supplier: BD
297252 297691
90004-954PK 51.87 USD
90004-954 90004-796 90004-956
BBL™ Sabouraud Brain Heart Infusion Agar, Slants, BD
Media for Microbiology Prepared Media for Microbiology
Sabouraud Brain Heart Infusion Agar Slants are used in qualitative procedures for the selective isolation and cultivation of pathogenic fungi from clinical and nonclinical specimens.
  • Peptones and infusions provide amino acids and nitrogen
  • Dextrose is an energy source
  • Sodium chloride provides essential electrolytes
  • Disodium phosphate buffers the medium to maintain the pH
  • Agar is the solidifying agent
Chloramphenicol is a broad-spectrum antibiotic that inhibits a wide range of gram-negative and gram-positive bacteria. Gentamicin is an aminoglycoside antibiotic that inhibits the growth of gram-negative bacteria.