Orange G solution, Surgipath® for cytology
Supplier: Leica Biosystems
|
Danger
|
Synonyms:
Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate
, Acid Orange 10
, Wool Orange 2G
, Orange G sodium salt
, CI 16230
Surgipath®
3801660
75810-356EA
183.78
USD
75810-356
Orange G solution, Surgipath® for cytology
Orange G
Orange G-6 is an alcohol-based stain used in the cytology laboratory to detect the presence of keratin in the cytoplasm.
- Alcohol-based
- Detects keratin in cytoplasm
It is manufactured by Leica Biosystems under the strictest controls using a standard formulation for conventional GYN/Non-GYN staining applications.
Formula:
C₁₆H₁₀N₂Na₂O₇S₂ MW: 452.38 g/mol |
MDL Number:
MFCD00012457 CAS Number: 1936-15-8 UN: 1987 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...