Surgipath® ActivFlo Biopsy II Cassettes, Leica
Supplier: Leica Biosystems
39LC-610-2-L 39LC-610-4-L 39LC-610-5-L 39LC-610-6-L 39LC-610-9-L
75809-464CS 543.95 USD
75809-464 75809-466 75809-468 75809-470 75809-472
Surgipath® ActivFlo Biopsy II Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
ActivFlo Biopsy II cassettes feature a patented “Fluidic Flow” pore design for maximum reagent exchange.
  • Available in 5 colors
  • Validated for Leica IP C Cassette Printer
  • <1% printing error rate
  • Two-piece, lids packaged separately
  • 45° writing surface
With a 0.67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes feature sloped walls, creating a basket for small tissue management.
Envase: Lids packaged separately.