Sodium dihydrogen phosphate hydrate ≥98% ACS, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Sodium phosphate monobasic, Sodium dihydrogen orthophosphate hydrate

567545-1KG 567545-500GM
80058-646EA 192.11 USD
80058-646 80058-650
Sodium dihydrogen phosphate hydrate ≥98% ACS, Calbiochem®
Sodium dihydrogen phosphate hydrate

Useful in conjunction with sodium phosphate, Dibasic in the preparation of biological buffers.


Special Instructions: Following reconstitution, store at room temperature. Stock solutions are stable for several months at room temperature

Formula: NaH₂PO₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5

Order Now

Specification Test Results

Form White Solid
Purity ≥98% by assay
Contaminants Heavy metals: ≤0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR