Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥99.5% (by HPLC)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

12831-25G 12831-1G
102571-268EA 667.63 USD
102571-268 CH12831-1G
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥99.5% (by HPLC)
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)

Sulfo-NHS is used for preparing hydrophilic active esters, e.g. for crosslinking agents.

Formula: C₄H₄NNaO₆S
Melting Pt: ≥ 360 ºC
Storage Temperature: Refrigerator
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Specification Test Results

Synonyms Hydroxy-2,5-dioxo-pyrrolidine-3-sulfonic-acid sodium salt  Sulfo-NHS
Purity Limit ≥99.5% (HPLC)
Appearance White to off-white powder
Storage Temperature Store at 0 - 8 °C
Comments Sulfo-NHS is used for preparing hydrophilic active esters, e.g. for crosslinking agents.
Solubility Clear, freely soluble in water
Infrared Spectrum Conforms to Structure
Analysis by Mass Spectra Conforms to Structure
1H NMR Conforms to Structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR