Iron(III) citrate

Supplier: CHEM IMPEX INTL INC MS

01445-1KG 01445-1KG
102573-636EA 173.75 USD
102573-636 CH01445-1KG
Iron(III) citrate
Iron(III) citrate
Formula: C₆H₅FeO₇
MW: 244.95
Storage Temperature: Ambient
CAS Number: 28633-45-6

Order Now

Specification Test Results

Appearance Orange-brown powder
Solubility (hot water) 1 g dissolves in 20 mL, gives a clear solution
Solubility 1 g dissolves in 100 mL water to give a clear, yellow to yellowish green solution
Chloride (Cl) ≤0.05%
Ammonium (NH₄⁺) ≤0.5%
Arsenic (As) ≤0.005%
Cadmium (Cd) ≤0.005%
Copper (Cu) ≤0.005%
Lead (Pb) ≤0.005%
Zinc (Zn) ≤0.005%
Ferrous (Fe²⁺) ≤5%
Water Content (Karl Fisher) ≤27%
Purity 18 - 20%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR