Anti-IgG Goat Antibody

Supplier: Equitech-Bio
GAHG80FC-0100 GAHG80FC-0500
103219-296EA 469.23 USD
103219-296 103219-298
Anti-IgG Goat Antibody
Antibodies
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR