Difco™ Sabouraud Dextrose Broth, BD
Supplier: BECTON DICKINSON DE MEXICO, S. MX
238210 238230
90003-092EA 531.03 USD
90003-092 90003-096
Difco™ Sabouraud Dextrose Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Broth is used for the cultivation of yeasts, molds, and aciduric microorganisms.
  • Peptone provides nitrogen, vitamins, minerals, amino acids, and growth factors
  • Low pH promotes the growth of fungi
  • Dehydrated appearance: Cream to yellow, free-flowing
The addition of antimicrobics further increases the selectivity of the medium.