Horse Albumin (from Serum)
Supplier: Equitech-Bio
ESA62-0001 ESA62-0010
103219-912EA 381.54 USD
103219-912 103219-914
Horse Albumin (from Serum)
Proteins and Peptides
Prepared from equine serum by heat-shock fractionation.
  • Purity: ≥ 96%
  • IgG level: not detected
Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Prepared from equine serum by heat-shock fractionation.