Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
Collected from 8-12 week-old sexually mature rats (Sprague-Dawley).
- Sterility tested
- Lot to lot consistency
- Low hemoglobin levels
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Subject to CITES regulations.