Feline Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SFPC35-0100
SFPC35-0500
SFPE35-0100
SFPE35-0500
SFPH35-0100
SFPH35-0500
FPC07-0100
FPC07-0500
FPE07-0100
FPE07-0500
FPH07-0100
FPH07-0500
103219-702EA
658.46
USD
103219-702
103219-704
103219-706
103219-708
103219-710
103219-712
103219-714
103219-716
103219-718
103219-720
103219-722
103219-724
Feline Plasma with Citrate, Equitech- Bio, Inc.
Plasma
Collected from sexually mature, mixed sex, mixed breed cats.
- Sterility tested
- Endotoxin and mycoplasma testing available
- Lot to lot consistency
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Plasma from specific breeds are available by request. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Mycoplasma testing available. Heat-inactivation is available if desired.