Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
CAPC07-0100 CAPC07-0500 CAPE07-0100 CAPE07-0500 CAPH07-0100 CAPH07-0500
103219-062EA 191.01 USD
103219-062 103219-064 103219-066 103219-684 103219-690 103219-692 103219-694 103219-696 103219-698 103219-700
Canine Plasma, Equitech- Bio, Inc.
Plasma
Collected from sexually mature, mixed sex, mixed breed dogs.
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Plasma from specific breeds are available by request. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Mycoplasma testing available. Heat-inactivation is available if desired.