Calf Serum, Equitech- Bio Inc.
Supplier: Equitech-Bio
SBCU30-0100 SBCU30-0500 SBCU32-0100 SBCU32-0500 BCSU05-0100 BCSU05-0500
103219-016EA 76.92 USD
103219-016 103219-018 103219-020 103219-022 103219-024 103219-026
Calf Serum, Equitech- Bio Inc.
Sera (Serum)
Collected from calves of U.S. origin
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Sterility tested and sterile filtered to 0.2 micron absolute. Low hemoglobin levels. Physiochemical constituents are measured to ensure lot to lot consistency. Endotoxin and mycoplasma testing is available while heat inactivation is available if desired.
Can be used in cell growth studies and immunoassays.