Hydrogen peroxide 3% USP antiseptic 10 Volume, Hydrox Laboratories
Supplier: Healthcare Products
HDXD0012
HDXD0011
75830-256CS
21.6
USD
75830-256
75830-254
Hydrogen peroxide 3% USP antiseptic 10 Volume, Hydrox Laboratories
Hydrogen peroxide
Formula:
H₂O₂ MW: 34.01 g/mol |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...