(3-Hydroxyoxetan-3-ylmethyl)carbamic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

115899-500MG 115899-1G 115899-5G
102898-688EA 1512.31 USD
102898-688 102898-690 102898-692
(3-Hydroxyoxetan-3-ylmethyl)carbamic acid tert-butyl ester ≥95%
(3-Hydroxyoxetan-3-ylmethyl)carbamic acid tert-butyl ester

Hazard: Irritant

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
MDL Number: MFCD22628739
CAS Number: 1408076-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR