cis-4-Aminotetrahydropyran-3-ol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (3R,4R)-4-Aminotetrahydro-2H-pyran-3-ol hydrochloride

116013-1G 116013-10G
102900-196EA 1787.69 USD
102900-196 102900-200
cis-4-Aminotetrahydropyran-3-ol hydrochloride ≥95%
cis-4-Aminotetrahydropyran-3-ol hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
MDL Number: MFCD28099084
CAS Number: 1523530-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR