[3-(Oxetan-3-yloxy)phenyl]methanamine ≥95%

Supplier: Matrix Scientific

115939-250MG 115939-1G
102899-918EA 963.08 USD
102899-918 102899-922
[3-(Oxetan-3-yloxy)phenyl]methanamine ≥95%
[3-(Oxetan-3-yloxy)phenyl]methanamine

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD21847942
CAS Number: 1416323-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR