3-(4-Nitrophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115927-250MG 115927-500MG 115927-100MG
102898-816EA 2792.31 USD
102898-816 102898-818 102899-024
3-(4-Nitrophenyl)oxetane-3-carboxylic acid ≥95%
3-(4-Nitrophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₅
MDL Number: MFCD22544049
CAS Number: 1393544-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR