3-(4-Methylphenyl)oxetane-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
115921-100MG
115921-250MG
115921-500MG
102898-814EA
1373.85
USD
102898-814
102898-996
102898-998
3-(4-Methylphenyl)oxetane-3-carboxylic acid ≥95%
3-(4-Methylphenyl)oxetane-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂O₃ |
MDL Number:
MFCD22628747 CAS Number: 1416323-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...